missing translation for 'onlineSavingsMsg'
Learn More

Olmesartan, MedChemExpress

Product Code. 16447226
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16447226 100 mg 100mg
16431826 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16447226 Supplier Medchem Express Supplier No. HY17004
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Olmesartan (RNH-6270) is an angiotensin II receptor (AT1R) antagonist used to treat high blood pressure.

Olmesartan (RNH-6270) is an angiotensin II receptor (AT1R) antagonist used to treat high blood pressure.

Specifications

Chemical Name or Material Olmesartan
CAS 144689-24-7
Color White
Purity Grade Notes Research
Molecular Formula C24H26N6O3
Quantity 100 mg
Synonym RNH-6270
Solubility Information DMSO : 50 mg/mL (111.98 mM; Need ultrasonic)
SMILES O=C(O)C1=C(N=C(N1CC2=CC=C(C=C2)C3=CC=CC=C3C4=NN=NN4)CCC)C(O)(C)C
Molecular Weight (g/mol) 446.5
Formula Weight 446.5
Percent Purity 98.2%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.