missing translation for 'onlineSavingsMsg'
Learn More

Olmesartan-d6 Acid (dimethyl-d6), CDN

Product Code. 30319899
Change view
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Product Code. Quantity unitSize
30319899 0.01 g 0.01g
30313311 0.005 g 0.01g
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30319899

Brand: CDN CDND75220.01G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Olmesartan-d6
CAS 1185144-74-4
Molecular Formula C24 D6 H20 N6 O3
Quantity 0.01 g
Synonym 1 H-Imidazole-5-carboxylic acid, 4-[1-hydroxy-1-(methyl-d3)ethyl-2,2,2-d3]-2-propyl-1-[[2'-(2 H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]- (ACI), 4-[1-Hydroxy-1-(methyl-d3)ethyl-2,2,2-d3]-2-propyl-1-[[2'-(2 H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1 H-imidazole-5-carboxylic acid (ACI), 5-(1,1,1,3,3,3-Hexadeuterio-2-hydroxypropan-2-yl)-2-propyl-3-[[4-[2-(2 H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazole-4-carboxylic acid, Olmesartan-d6, Olmesartan acid-d6, Olmesartan-d6 Acid (dimethyl-d6), Olmesartan-D6 (1-(methyl-D3)ethyl-2,2,2-D3)
InChI Formula InChI=1 S/C24H26N6O3/c1-4-7-19-25-21(24(2,3)33)20(23(31)32)30(19)14-15-10-12-16(13-11-15)17-8-5-6-9-18(17)22-26-28-29-27-22/h5-6,8-13,33 H,4,7,14H2,1-3H3,(H,31,32)(H,26,27,28,29)/i2D3,3D3
SMILES [2 H]C([2 H])([2 H])C(O)(c1nc(CCC)n(Cc2ccc(cc2)c3ccccc3c4nnn[nH]4)c1C(=O)O)C([2 H])([2 H])[2 H]
IUPAC Name 5-(1,1,1,3,3,3-hexadeuterio-2-hydroxypropan-2-yl)-2-propyl-3-[[4-[2-(1 H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazole-4-carboxylic acid
Molecular Weight (g/mol) 452.539
Formula Weight 452.244 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.