missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olivetol, MedChemExpress
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Olivetol is a naturally phenol found in lichens and produced by certain insects, acting as a competitive inhibitor of the cannabinoid receptors CB1 and CB2. Olivetol also inhibits CYP2C19 and CYP2D6 activity, with IC50s of 15.3 μM, 7.21 μM and Kis of 2.71 μM, 2.87 μM, respectively.
Specifications
Specifications
| Chemical Name or Material | Olivetol |
| CAS | 500-66-3 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C11H16O2 |
| Quantity | 100 mg |
| Solubility Information | DMSO : 100 mg/mL (554.82 mM; Need ultrasonic) |
| SMILES | OC1=CC(CCCCC)=CC(O)=C1 |
| Molecular Weight (g/mol) | 180.24 |
| Formula Weight | 180.24 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction