missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olive Oil, TRC
Specifications
Specifications
| Chemical Name or Material | Olive Oil |
| CAS | 8001-25-0 |
| Quantity | 500 g |
| Synonym | Olive oil, Cropure OL, Cropure oil, Fats and Glyceridic oils','olive, Giant, Oils','glyceridic','olive, Oils','olive, Olea europaea ext., Olea europaea oil, Oleum Olivarum, Oliterra Pure Olive Oil, Olive','ext., Sulfur olive oil |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?