missing translation for 'onlineSavingsMsg'
Learn More

Olive oil, pure, virgin

Product Code. 10566982 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10566982 500 mL Glass bottle 500mL
10667031 25 mL Glass bottle 25mL
11964041 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10566982 Supplier Thermo Scientific Acros Supplier No. 416545000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 8001-25-0

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8001-25-0

Specifications

Acidity 2.0mg KOH/g max.
Absorbance 0.2 (270nm, 1g/100ml cyclohexane) max. at 425nm
Color Yellow
Density 0.9135g/mL
Flash Point 225°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4670 to 1.4710
Quantity 500 mL
Saponification Value 184 to 196mg KOH/g
Merck Index 14,6836
Specific Gravity 0.9135
Solubility Information Solubility in water: .. Other solubilities: practically insoluble in alcohol, miscible with light petroleum (50-60°C)
Grade Pure
Concentration or Composition (by Analyte or Components) of fatty acids: 0.1 % max. (< C16) 7.5 to 20.0 % (Palmitic acid) 3.5 % max. (Palmitoleic acid) 0.5 to 5.0 % (Stearic acid) 56.0 to 85.0 % (Oleic acid) 3.5 to 20.0 % (Linoleic acid) 1.2 % max. (Linolenic acid) 0.7 % max. (Arachidic acid) 0.4 % max. (Eic
Physical Form Powder
Chemical Name or Material Olive oil
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.