missing translation for 'onlineSavingsMsg'
Learn More

Olive oil, pure, refined

Product Code. 10370674 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10370674 2.5 L Glass bottle 2.5 L
10556982 25 mL Glass bottle 25mL
10494121 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10370674 Supplier Thermo Scientific Acros Supplier No. 416530025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 8001-25-0

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8001-25-0
MDL Number MFCD00131764

Specifications

Acidity 0.3 max. (mg KOH/g)
Absorbance 1.20 max. (1g/100ml cyclohexane) measured at the maximum between 264nm and 276nm
Color Colorless or Green-Yellow
Density 0.9135g/mL
Flash Point 225°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4670 to 1.4710
Quantity 2.5 L
Merck Index 14, 6836
Specific Gravity 0.9135
Solubility Information Solubility in water: .. Other solubilities: practically insoluble in alcohol, miscible with light petroleum (50-60°C)
Grade Pure
Concentration or Composition (by Analyte or Components) of fatty acids: 0.1% max. (< C16), of the sterol fraction: Sterols: min. 93.0%, Cholesterol: max. 0.5%, Campesterol: max. 4.0%, delta 7-Stigmasterol: max. 0.5%
Physical Form Viscous Liquid
Chemical Name or Material Olive oil
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.