missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Oligomycin A

Product Code. 18045222
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18045222 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18045222 Supplier Tocris Bioscience™ Supplier No. 4110/5
  • ONLINE EXCLUSIVE PRICE
    £173.00
    £162.75 / 5mg
    Save £10.25
    5% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibitor of mitochondrial ATPase

Oligomycin A is an inhibitor of mitochondrial ATP synthase and uncoupler of oxidative phosphorylation. Antibiotic; exhibits anti-tumor activity.
TRUSTED_SUSTAINABILITY

Specifications

CAS 579-13-5
Quantity 5 mg
Molecular Formula C45H74O11
Formula Weight 791.06
Product Type Oligomycin A
Purity 98%
Molecular Weight (g/mol) 791.08
Chemical Name or Material Oligomycin A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.