Learn More
Invitrogen™ Oligodendrocyte Marker O1 Monoclonal Antibody (O1), eFluor™ 660, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£198.00 - £402.00
Specifications
| Antigen | Oligodendrocyte Marker O1 |
|---|---|
| Clone | O1 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15540007
|
Invitrogen™
50-6506-80 |
25 μg |
£198.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15550007
![]() |
Invitrogen™
50-6506-82 |
100 μg |
£402.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: This O1 monoclonal antibody reacts with galactocerebroside and other lipids found on late oligodendrocyte precursors. O1 has been reported to react to all species tested (rat, mouse, human, chicken). Oligodendrocytes function as myelin-producing cells within the central nervous system ensheathing the axons of neurons with myelin in order to facilitate rapid axonal conduction. O1 and O4 monoclonal antibodies serve as stage-specific markers of oligodendrocyte development with O4 marking early oligodendrocyte progenitors and O1-labeling late oligodendrocyte progenitors. O4-positive cells differentiate to become O1-positive, which can in turn become mature oligodendrocytes. Applications Reported: This O1 antibody has been reported for use in flow cytometric analysis, immunohistochemical staining of frozen tissue sections, and immunocytochemistry. Applications Tested: This O1 antibody has been tested by immunocytochemistry on fixed, differentiated OLN93 cells and can be used at less than or equal to 20 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Oligodendrocytes function as myelin-producing cells within the central nervous system ensheathing the axons of neurons with myelin in order to facilitate rapid axonal conduction. O1 and O4 monoclonal antibodies serve as stage-specific markers of oligodendrocyte development with O4 marking early oligodendrocyte progenitors and O1-labeling late oligodendrocyte progenitors. O4-positive cells differentiate to become O1-positive, which can in turn become mature oligodendrocytes.Specifications
| Oligodendrocyte Marker O1 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| IgM | |
| Affinity chromatography | |
| Antibody |
| O1 | |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry | |
| eFluor 660 | |
| Mouse | |
| Human, Mouse, Rat, Chicken | |
| O1 Marker | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.