Learn More
Thermo Scientific™ OLIGO™ R3 Reversed Phase Columns
Shop All Applied Biosystems ProductsDescription
Oligo™ R3 bulk resin is a rigid polymeric bead designed for reversed-phase chromatography of polynucleotides and other biomolecules. The combination of 30-μm particle size, resistance to high pH, and high-phase ratio, makes Oligo R3 resin particularly suited to preparative purification of synthetic oligonucleotides and peptides. The packing consists of cross-linked poly-divinylbenzene particles.
Oligo R3 bulk resin differs somewhat in selectivity from conventional silica-based, reversed-phase resins and is similar to high-carbon-loading through C18 supports. In addition, the complete lack of residual silanol activity may modify the selectivity of ionic species.
Features of Oligo R3 resin and pre-packed columns include:
- High performance chromatography resin for analytical to process scale separations
- High resolution preparative purification at high flow rates
- Chemical stability—allows aggressive cleaning and sanitization
- Poly-divinylbenzene particles—yield a robust, easily packable matrix
- Combines low back pressure, high pH stability, and high capacity
Specifications
Specifications
| Column Format | Prepacked Column |
| Diameter (Metric) | 4.6 mm |
| Length (Metric) | 100 mm |
| Max. Pressure | 2400 psi (170 bar) |
| Particle Size | 30 μm |
| Pore Size | 350 Å |
| Quantity | 1 Column |
| Stationary Phase | Phenyl |
| Content And Storage | Store at room temperature. |
| Product Line | POROS |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.