missing translation for 'onlineSavingsMsg'
Learn More

Olig3 Antibody (257914) [DyLight 650], Novus Biologicals™

Product Code. 30737498 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737498 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737498 Supplier Novus Biologicals Supplier No. FAB24561W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Olig3 Monoclonal antibody specifically detects Olig3 in Human,Mouse samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Olig3
Applications Immunohistochemistry
Classification Monoclonal
Clone 257914
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias Bhlhb7, BHLHE20, bHLHe20oligo3, Class B basic helix-loop-helix protein 7, Class E basic helix-loop-helix protein 20, Oligo3, oligodendrocyte transcription factor 3
Host Species Mouse
Immunogen E. coli-derived recombinant human Olig3, Met1-Lys272, Accession # Q7RTU3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 167826
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.