missing translation for 'onlineSavingsMsg'
Learn More

Olig2 Antibody (OLIG2/6695R), DyLight 350, Novus Biologicals™

Product Code. 18380872 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18380872 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18380872 Supplier Novus Biologicals Supplier No. NBP314217UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Olig2 Monoclonal antibody specifically detects Olig2 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Olig2
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OLIG2/6695R
Conjugate DyLight 350
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias BHLHB1, BHLHE19, Class B basic helix-loop-helix protein 1, Class E basic helix-loop-helix protein 19, human protein kinase C-binding protein RACK17, OLIGO2, oligodendrocyte lineage transcription factor 2, oligodendrocyte transcription factor 2, oligodendrocyte-specific bHLH transcription factor 2, protein kinase C binding protein 2, Protein kinase C-binding protein 2, Protein kinase C-binding protein RACK17, RACK17helix-loop-helix protein, class B, 1
Host Species Rabbit
Immunogen Recombinant fragment (around aa200-300) of human Olig2 protein (exact sequence is proprietary) (Uniprot: Q13516)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Chromatin Research, Glia Markers, Neuroscience, Oligodendrocyte Cell Markers, Stem Cell Markers, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 10215
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.