missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OLIG1 Recombinant Rabbit Monoclonal Antibody (23GB2350), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | OLIG1 |
|---|---|
| Clone | 23GB2350 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087470
![]() |
Invitrogen™
MA552504 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088517
|
Invitrogen™
MA552505 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OLIG1 Recombinant Monoclonal antibody specifically detects OLIG1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| OLIG1 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q8TAK6, Q9JKN5, Q9WUQ3 | |
| 116448, 50914, 60394 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2350 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AI836478; AW494459; basic domain, helix-loop-helix protein, class B, 6; BHLHB6; bHLHb7; bHLHe20; bHLHe21; class B basic helix-loop-helix protein 6; Class E basic helix-loop-helix protein 21; LOW QUALITY PROTEIN: oligodendrocyte transcription factor 1; Olg1; Olg-1; Olg-1 bHLH protein; OLIG1; olig2; olig3; Oligo1; Oligo2 antibody; oligodendrocyte lineage transcription factor 1; oligodendrocyte transcription factor 1; Oligodendrocyte-specific bHLH transcription factor 1 | |
| A synthesized peptide derived from human Olig1 (200-271AA). | |
| Primary | |
| -20°C | |
| OLIG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title