missing translation for 'onlineSavingsMsg'
Learn More

Olig1 Antibody (257219) [mFluor Violet 450 SE], Novus Biologicals™

Product Code. 30737281 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737281 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737281 Supplier Novus Biologicals Supplier No. FAB2417MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Olig1 Monoclonal antibody specifically detects Olig1 in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Olig1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 257219
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias bHLHb6, BHLHE21, Class B basic helix-loop-helix protein 6, Class E basic helix-loop-helix protein 21, helix-loop-helix protein, class B, 6, oligo1, oligodendrocyte lineage transcription factor 1, oligodendrocyte transcription factor 1, oligodendrocyte-specific bHLH transcription factor 1
Host Species Mouse
Immunogen E. coli-derived recombinant human Olig1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 116448
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.