missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OLFML3 Monoclonal Antibody (740404)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA561802

Product Code. 30534186

  • £357.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.

This gene encodes a member of the olfactomedin-like gene family which also includes genes encoding noelin, tiarin, myocilin, amassin, optimedin, photomedin, and latrophilin. The encoded protein is a secreted extracellular matrix glycoprotein with a C-terminal olfactomedin domain that facilitates protein-protein interactions, cell adhesion, and intercellular interactions. It serves as both a scaffold protein that recruits bone morphogenetic protein 1 to its substrate chordin, and as a vascular tissue remodeler with pro-angiogenic properties. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Feb 2017].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

OLFML3
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q8BK62
OLFML3
Mouse myeloma cell line NS0-derived recombinant mouse OLFM-L3, Gln22-Val406.
100 μg
Primary
Mouse
Antibody
IgG2b
Western Blot
740404
Unconjugated
OLFML3
2810002E22Rik; AI115209; AW550633; HNOEL-iso; hOLF44; mONT3; OLF44; olfactomedin like 3; olfactomedin-like 3; olfactomedin-like protein 3; OLFML3; ONT3; PSEC0035; PSEC0173; PSEC0244; UNQ663/PRO1294
Rat
Protein A/G
RUO
99543
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.