missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OLFML2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113006
This item is not returnable.
View return policy
Description
OLFML2B Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes an olfactomedin domain-containing protein. Most olfactomedin domain-containing proteins are secreted glycoproteins.
Specifications
| OLFML2B | |
| Polyclonal | |
| Unconjugated | |
| OLFML2B | |
| 1110018N05Rik; 4832415H08Rik; AI467542; olfactomedin like 2B; olfactomedin-like 2B; olfactomedin-like protein 2B; Olfml2b; Photomedin-2 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 25903, 320078 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q3V1G4, Q68BL8 | |
| OLFML2B | |
| Recombinant Human Olfactomedin-like protein 2B protein (451-750AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction