missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OLFML2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554577
This item is not returnable.
View return policy
Description
Immunogen sequence: FYTVETVSSG TDCRCSCTAP PSSLNPCENE WKMEKLKKQA PELLKLQSMV DLLEGTLYSM DLMKVHAYVH KVASQMNTLE ESIKANLSRE N Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 92%.
OLFML2A contains one olfactomedin-like domain. Its exact function is unknown. There are three named isoforms produced by alternative splicing.
Specifications
| OLFML2A | |
| Polyclonal | |
| Unconjugated | |
| OLFML2A | |
| 4932431K08Rik; mFLJ00237; olfactomedin like 2A; olfactomedin-like 2A; olfactomedin-like protein 2A; OLFML2A; Photomedin-1; PRO34319; UNQ9394/PRO34319 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 169611 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q68BL7 | |
| OLFML2A | |
| Recombinant protein corresponding to Human OLFML2A. Recombinant protein control fragment (Product #RP-93076). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction