missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OLFM4 Recombinant Rabbit Monoclonal Antibody (24GB680)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557529
This item is not returnable.
View return policy
Description
OLFM4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
OLFM4 is a 510 amino acid extracellular matrix glycoprotein belonging to the olfactomedin-related protein family and found to be selectively expressed in inflammed colonic epithelium. It has one signal peptide, six N-linked glycosylation motif and forms disulfide-bonded multimers. OLFM4 a transcription factor may have a matrix-related function in differentiation. Constitutively expressed OLFM4 in cancer cells binds to the potent apoptosis inducer GRIM-19 and has a novel antiapoptotic action and hence might serve as a diagnostic marker for human cancers. Reports suggest that OLFM4 promotes proliferation of PANC-1 cells by favoring transition from the S to G2/M phase. Potential interaction of OLFM4 with endogenous cell surface lectins and cadherins may mediate cell adhesion.
Specifications
| OLFM4 | |
| Recombinant Monoclonal | |
| 1.05 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q3UZZ4, Q6UX06 | |
| OLFM4 | |
| A synthesized peptide derived from human OLFM4 (aa 100-157). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB680 | |
| Unconjugated | |
| OLFM4 | |
| Antiapoptotic protein GW112; bA209J19.1; GC1; G-CSF-stimulated clone 1 protein; Gm296; Gm913; GW112; hGC-1; hOLfD; olfactomedin 4; olfactomedin-4; OlfD; OLFM4; OLM4; pPD4; PU.1 difference product 4; UNQ362; UNQ362/PRO698 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10562, 290409, 380924 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction