missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OLFM4 Antibody (12), Novus Biologicals™
Mouse Monoclonal Antibody
£180.00 - £256.00
Specifications
| Antigen | OLFM4 |
|---|---|
| Clone | 12 |
| Dilution | Flow Cytometry 1:25-1:100, ELISA 1:1000-1:2000 |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18662097
|
Novus Biologicals
NBP3-06415-100ul |
100 μg |
£256.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18660057
|
Novus Biologicals
NBP3-06415-50ul |
50 μg |
£180.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OLFM4 Monoclonal antibody specifically detects OLFM4 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
| OLFM4 | |
| Flow Cytometry 1:25-1:100, ELISA 1:1000-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Antiapoptotic protein GW112, bA209J19.1, GC1, G-CSF-stimulated clone 1 protein, GW112hOLfD, hGC-1, KIAA4294, olfactomedin 4, olfactomedin-4, OlfD, OLM4, UNQ362 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human OLFM4 (Accession#: NP_006409.3; Met 1-Gln 510). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 12 | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| 0.2 um filtered solution in PBS | |
| 10562 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title