missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OLFM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565590
This item is not returnable.
View return policy
Description
Immunogen sequence: SLFYNKYQSNV VVKYHFRSRS VLVQRSLPGA GYNNTFPYSW GGFSDMDFMV DES Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.
Involved in transforming growth factor beta (TGF-beta)-induced smooth muscle differentiation. TGF-beta induces expression and nuclear translocation of OLFM2 where it binds to SRF, causing its dissociation from the transcriptional repressor HEY2/HERP1 and facilitating binding of SRF to target genes. Plays a role in AMPAR complex organization.
Specifications
| OLFM2 | |
| Polyclonal | |
| Unconjugated | |
| OLFM2 | |
| A030009A06Rik; neuronal olfactomedin related ER localized protein 2; NOE2; noelin 2; NOELIN2; Noelin-2; NOELIN2_V1; olfactomedin 2; olfactomedin-2; OlfC; OLFM2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 93145 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95897 | |
| OLFM2 | |
| Recombinant Human OLFM2. Recombinant protein control fragment (Product #RP-98972). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction