missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OLFM-L3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68601-25ul
This item is not returnable.
View return policy
Description
OLFM-L3 Polyclonal antibody specifically detects OLFM-L3 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| OLFM-L3 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| HNOEL-isoOLF44, hOLF44, olfactomedin-like 3, olfactomedin-like protein 3 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LVEYMERRLAALEERLAQCQDQSSRHAAELRDFKNKMLPLLEVAEKEREALRTEADTISGRVDRLEREVDYLETQNPALPCVEFDEKVTGGPGTKGKGRRNE | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 56944 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction