missing translation for 'onlineSavingsMsg'
Learn More

Olfactory Receptor OR2A4 Antibody, Novus Biologicals™

Product Code. 18613016 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18613016 0.1 mL 0.1mL
18686565 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18613016 Supplier Novus Biologicals Supplier No. NBP246848

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Olfactory Receptor OR2A4 Polyclonal antibody specifically detects Olfactory Receptor OR2A4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Olfactory Receptor OR2A4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2), 40% Glycerol
Gene Accession No. O95047
Gene Alias Olfactory receptor 2A10, olfactory receptor 2A4, Olfactory receptor OR6-37, olfactory receptor, family 2, subfamily A, member 10, olfactory receptor, family 2, subfamily A, member 4, OR2A10
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: RMLVNLLHPAKPISFAGRMM
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Olfactory
Primary or Secondary Primary
Gene ID (Entrez) 79541
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.