missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olfactory Receptor OR2A4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | Olfactory Receptor OR2A4 |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18686565
|
Novus Biologicals
NBP2-46848-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18613016
|
Novus Biologicals
NBP2-46848 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Olfactory Receptor OR2A4 Polyclonal antibody specifically detects Olfactory Receptor OR2A4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| Olfactory Receptor OR2A4 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Olfactory | |
| PBS (pH 7.2), 40% Glycerol | |
| Olfactory receptor 2A10, olfactory receptor 2A4, Olfactory receptor OR6-37, olfactory receptor, family 2, subfamily A, member 10, olfactory receptor, family 2, subfamily A, member 4, OR2A10 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: RMLVNLLHPAKPISFAGRMM | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| O95047 | |
| 79541 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title