missing translation for 'onlineSavingsMsg'
Learn More

Olfactomedin-1/Noelin-1 Antibody, Novus Biologicals™

Product Code. 18354493 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354493 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18354493 Supplier Novus Biologicals Supplier No. NBP312484

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Olfactomedin-1/Noelin-1 Polyclonal antibody specifically detects Olfactomedin-1/Noelin-1 in Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Olfactomedin-1/Noelin-1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:10000
Formulation Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide
Gene Alias AMYolfactomedin related ER localized protein, neuroblastoma protein, Neuronal olfactomedin-related ER localized protein, NOE1noelin, NOEL1, NOELIN, NOELIN1, OlfA, olfactomedin 1, olfactomedin-1, pancortin 1
Host Species Rabbit
Immunogen N-epitope synthetic peptide taken within amino acid region 1-60 on Noelin-1 protein.
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10439
Target Species Mouse
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.