missing translation for 'onlineSavingsMsg'
Learn More

Olfactomedin-1/Noelin-1 Antibody (2E9-1A2), Novus Biologicals™

Product Code. 18370139 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370139 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18370139 Supplier Novus Biologicals Supplier No. H00010439M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Olfactomedin-1/Noelin-1 Monoclonal antibody specifically detects Olfactomedin-1/Noelin-1 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Olfactomedin-1/Noelin-1
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2E9-1A2
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH00189
Gene Alias AMYolfactomedin related ER localized protein, neuroblastoma protein, Neuronal olfactomedin-related ER localized protein, NOE1noelin, NOEL1, NOELIN, NOELIN1, OlfA, olfactomedin 1, olfactomedin-1, pancortin 1
Host Species Mouse
Immunogen OLFM1 (AAH00189, 1 a.a. ~ 169 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MPGRWRWQRDMHPARKLLSLLFLILMGTELTQNKRENKAEKMGGPESERKTTGEKTLNELPLFCLEAHAGSLALPRMCSPNPNPAVGLCRPAYPQSPSPGAAQTISQSLLERFCMASRREVFLAPGRPGGGWWLCTVAGQMHSFMCTHTHTHAHTGEQIPAEKSQPGPD
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 10439
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.