missing translation for 'onlineSavingsMsg'
Learn More

Oleyl alcohol, tech. 80-85%, Thermo Scientific Chemicals

Product Code. 11459156
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459156 500 g 500g
11449156 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459156 Supplier Thermo Scientific Alfa Aesar Supplier No. A18018.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Oleyl alcohol is used in softening and lubrication of textile fabrics, and in the production of carbon paper, stencil paper, and printing ink. It finds application in cosmetic products viz skin creams and lotions as a thickner, hair conditioners and hair coating shampoos. It is utilized as an antifoaming agent and cutting lubricant, as the precursor for the preparation of its sulfuric ester derivatives, which are used in detergents and wetting agents. It plays a vital role in various formulations for drug delivery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oleyl alcohol is used in softening and lubrication of textile fabrics, and in the production of carbon paper, stencil paper, and printing ink. It finds application in cosmetic products viz skin creams and lotions as a thickner, hair conditioners and hair coating shampoos. It is utilized as an antifoaming agent and cutting lubricant, as the precursor for the preparation of its sulfuric ester derivatives, which are used in detergents and wetting agents. It plays a vital role in various formulations for drug delivery.

Solubility
Miscible with alcohol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.

Notes
Incompatible with oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143-28-2
Molecular Formula C18H36O
Molecular Weight (g/mol) 268.49
MDL Number MFCD00002993
InChI Key ALSTYHKOOCGGFT-KTKRTIGZSA-N
Synonym oleyl alcohol, cis-9-octadecen-1-ol, z-octadec-9-en-1-ol, ocenol, conditioner 1, dermaffine, lancol, novol, oceol, oleol
PubChem CID 5284499
ChEBI CHEBI:73504
IUPAC Name (9Z)-octadec-9-en-1-ol
SMILES CCCCCCCC\C=C/CCCCCCCCO

Specifications

Melting Point ∽6°C
Density 0.849
Boiling Point 206°C to 207°C (13 mm)
Flash Point 180°C (356°F)
Assay Percent Range 0.85
Refractive Index 1.46
Quantity 500 g
Beilstein 1723962
Solubility Information Miscible with alcohol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.
Formula Weight 268.48
Grade Technical
Physical Form Liquid
Chemical Name or Material Oleyl alcohol
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.