missing translation for 'onlineSavingsMsg'
Learn More

Oleoylethanolamide, MedChemExpress

Product Code. p-200096256
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16446847

Brand: Medchem Express HY107542

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Oleoylethanolamide is a high affinity endogenous PPAR-α agonist, which plays an important role in the treatment of obesity and arteriosclerosis.

Oleoylethanolamide is a high affinity endogenous PPAR-α agonist, which plays an important role in the treatment of obesity and arteriosclerosis.

Specifications

Chemical Name or Material Oleoylethanolamide
Physical Form Solid
CAS 111-58-0
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C20H39NO2
For Use With (Application) COVID-19-anti-virus
Synonym N-Oleoylethanolamide Oleamide MEA Oleic acid monoethanolamide
Solubility Information DMSO : 20.83 mg/mL (63.99 mM; Need ultrasonic)
SMILES CCCCCCCC/C=C\CCCCCCCC(NCCO)=O
Molecular Weight (g/mol) 325.53
Formula Weight 325.53
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.