missing translation for 'onlineSavingsMsg'
Learn More

Oleoylethanolamide, MedChemExpress

Product Code. 16446847
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16446847 5 mg 5mg
16404148 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16446847 Supplier Medchem Express Supplier No. HY107542
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Oleoylethanolamide is a high affinity endogenous PPAR-α agonist, which plays an important role in the treatment of obesity and arteriosclerosis.

Oleoylethanolamide is a high affinity endogenous PPAR-α agonist, which plays an important role in the treatment of obesity and arteriosclerosis.

Specifications

Chemical Name or Material Oleoylethanolamide
Physical Form Solid
CAS 111-58-0
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C20H39NO2
For Use With (Application) COVID-19-anti-virus
Synonym N-Oleoylethanolamide Oleamide MEA Oleic acid monoethanolamide
Solubility Information DMSO : 20.83 mg/mL (63.99 mM; Need ultrasonic)
SMILES CCCCCCCC/C=C\CCCCCCCC(NCCO)=O
Molecular Weight (g/mol) 325.53
Formula Weight 325.53
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.