missing translation for 'onlineSavingsMsg'
Learn More

Oleic Acid, Pure, Technical, Fisher Chemical™

Product Code. 12636747 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
25 L
Packaging:
Amber glass bottle
Plastic container
Unit Size:
2.5 L
25 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12636747 25 L Plastic container 25 L
10578810 2.5 L Amber glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12636747 Supplier Fisher Chemical Supplier No. O/0150/25

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 112-80-1
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.47
MDL Number MFCD00064242
InChI Key ZQPPMHVWECSIRJ-MDZDMXLPSA-N
Synonym oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid
PubChem CID 445639
ChEBI CHEBI:16196
IUPAC Name (Z)-octadec-9-enoic acid
SMILES CCCCCCCC\C=C\CCCCCCCC(O)=O

Specifications

Melting Point 13°C
Color Colorless
Boiling Point 360°C
Packaging Plastic container
Quantity 25 L
Vapor Density 9.7
Viscosity 39.1 mPaS at 20°C
Formula Weight 282.46g/mol
Vapor Pressure 1mmHg 176
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.