Learn More
Oleic Acid, Extra Pure, SLR, Fisher Chemical™
C18H34O2, CAS Number-112-80-1, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, glycon ro, glycon wo, oleate, oleic acid, pamolyn 100, wecoline oo, z-octadec-9-enoic acid, 2.5L, 360 deg.C, CHEBI:16196, Colorless, 282.46g/mol, ZQPPMHVWECSIRJ-KTKRTIGZSA-N, (Z)-octadec-9-enoic acid, 13 deg.C
Brand: Fisher Chemical O/0200/17
Code : 21
Additional Details : CAS Number : 112-80-1 Weight : 2.75000kg
Description
C18H34O2, CAS Number-112-80-1, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, glycon ro, glycon wo, oleate, oleic acid, pamolyn 100, wecoline oo, z-octadec-9-enoic acid, 2.5L, 360 deg.C, CHEBI:16196, Colorless, 282.46g/mol, ZQPPMHVWECSIRJ-KTKRTIGZSA-N, (Z)-octadec-9-enoic acid, 13 deg.C
Specifications
112-80-1 | |
13°C | |
C18H34O2 | |
Amber glass bottle | |
2.5L | |
CCCCCCCC\C=C\CCCCCCCC(O)=O | |
282.47 | |
CHEBI:16196 | |
Liquid | |
39.1 mPaS at 20°C |
Colorless | |
360°C | |
MFCD00064242 | |
oleic acid, cis-9-octadecenoic acid, cis-oleic acid, elaidoic acid, oleate, glycon wo, wecoline oo, pamolyn 100, glycon ro, z-octadec-9-enoic acid | |
ZQPPMHVWECSIRJ-MDZDMXLPSA-N | |
(Z)-octadec-9-enoic acid | |
445639 | |
282.46g/mol | |
9.7 | |
1mmHg 176 |
Safety and Handling
- Oleic acid
Signal Word
- Warning
Hazard Category
- Specific target organ toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P332+P313-If skin irritation occurs: Get medical advice/attention.