missing translation for 'onlineSavingsMsg'
Learn More

Oleanolic Acid, MedChemExpress

Product Code. 16479006
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479006 100 mg 100mg
16442356 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16479006 Supplier Medchem Express Supplier No. HYN0156
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Oleanolic acid (Caryophyllin) is a natural compound from plants with anti-tumor activities.

Oleanolic acid (Caryophyllin) is a natural compound from plants with anti-tumor activities.

Specifications

Chemical Name or Material Oleanolic Acid
CAS 508-02-1
Color White
Purity Grade Notes Research
Molecular Formula C30H48O3
Quantity 100 mg
Synonym Oleanic acid Caryophyllin
Solubility Information DMSO : 5 mg/mL (10.95 mM; Need ultrasonic)
SMILES C[C@]([C@@]1([H])[C@]2(CC[C@@H]3O)C)(CC[C@@]2([H])C3(C)C)[C@@](C4=CC1)(CC[C@]5(C(O)=O)[C@@]4([H])CC(C)(C)CC5)C
Molecular Weight (g/mol) 456.7
Formula Weight 456.7
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.