missing translation for 'onlineSavingsMsg'
Learn More

Oleandomycin, MedChemExpress

Product Code. 16474858
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16474858 10 mg 10mg
16467607 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16474858 Supplier Medchem Express Supplier No. HY116010
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Oleandomycin is a macrolide antibiotic structurally closely related to Erythromycin. Oleandomycin is similar to Erythromycin with antimicrobial activity.

Oleandomycin is a macrolide antibiotic structurally closely related to Erythromycin. Oleandomycin is similar to Erythromycin with antimicrobial activity.

Specifications

Chemical Name or Material Oleandomycin
Physical Form Powder
CAS 3922-90-5
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C35H61NO12
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 100 mg/mL (145.38 mM; Need ultrasonic)
SMILES O=C1[C@H](C)[C@@H](O)[C@@H](C)[C@@H](C)OC([C@H](C)[C@@H](O[C@]2([H])O[C@@H](C)[C@H](O)[C@@H](OC)C2)[C@H](C)[C@@H](O[C@@]3([H])[C@H](O)[C@@H](N(C)C)C[C@@H](C)O3)[C@@H](C)C[C@@]14CO4)=O
Molecular Weight (g/mol) 687.86
Formula Weight 687.86
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.