missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oleamide, Tocris Bioscience™
Sleep-inducing brain lipid
Chemical Identifiers
CAS | 301-02-0 |
---|---|
Molecular Formula | C18H35NO |
Molecular Weight (g/mol) | 281.484 |
InChI Key | FATBGEAMYMYZAF-KTKRTIGZSA-N |
Synonym | oleamide, oleic acid amide, oleylamide, oleyl amide, adogen 73, crodamide o, slip-eze, armoslip cp, crodamide or, cis-9,10-octadecenoamide |
PubChem CID | 5283387 |
ChEBI | CHEBI:116314 |
IUPAC Name | (Z)-octadec-9-enamide |
SMILES | CCCCCCCCC=CCCCCCCCC(=O)N |
Description
Endogenous sleep-inducing lipid. Acts as an agonist at the CB1 cannabinoid receptor (EC50 = 1.64 μM). Also appears to potentiate the actions of 5-HT on 5-HT2A and 2C receptors, and act via an allosteric regulatory site on 5-HT7 receptors.Chemical Identifiers
301-02-0 | |
281.484 | |
oleamide, oleic acid amide, oleylamide, oleyl amide, adogen 73, crodamide o, slip-eze, armoslip cp, crodamide or, cis-9,10-octadecenoamide | |
CHEBI:116314 | |
CCCCCCCCC=CCCCCCCCC(=O)N |
C18H35NO | |
FATBGEAMYMYZAF-KTKRTIGZSA-N | |
5283387 | |
(Z)-octadec-9-enamide |
Specifications
301-02-0 | |
oleamide, oleic acid amide, oleylamide, oleyl amide, adogen 73, crodamide o, slip-eze, armoslip cp, crodamide or, cis-9,10-octadecenoamide | |
CCCCCCCCC=CCCCCCCCC(=O)N | |
281.484 | |
CHEBI:116314 | |
>98% |
C18H35NO | |
FATBGEAMYMYZAF-KTKRTIGZSA-N | |
(Z)-octadec-9-enamide | |
5283387 | |
281.48 | |
Oleamide |
Safety and Handling
Recommended Storage : Store at +4°C