missing translation for 'onlineSavingsMsg'
Learn More

Oleamide, MedChemExpress

Product Code. 16494937
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494937 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16494937 Supplier Medchem Express Supplier No. HYN2327
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Oleamide is an endogenous fatty acid amide which can be synthesized de novo in the mammalian nervous system, and has been detected in human plasma.

Oleamide is an endogenous fatty acid amide which can be synthesized de novo in the mammalian nervous system, and has been detected in human plasma.

Specifications

Chemical Name or Material Oleamide
CAS 301-02-0
Color White
Purity Grade Notes Research
Molecular Formula C18H35NO
Quantity 50 mg
Solubility Information Ethanol : 25 mg/mL (88.82 mM; Need ultrasonic) ∣DMSO : 25 mg/mL (88.82 mM; ultrasonic and warming and heat to 60°C)
SMILES CCCCCCCC/C=C\CCCCCCCC(N)=O
Molecular Weight (g/mol) 281.48
Formula Weight 281.48
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.