missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ OLDA
GREENER_CHOICE

Product Code. 18005732
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18005732

Brand: Tocris Bioscience™ 1641/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent, selective endogenous TRPV1 agonist

Potent endogenous vanilloid TRPV1 (VR1) receptor agonist (EC50 = 36 nM at hVR1) with low affinity for rCB1 receptors (Ki = 1.6 μM). Potently induces VR1-mediated thermal hyperalgesia in rats in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105955-11-1
Molecular Formula C26H43NO3

Specifications

Quantity 5 mg
Formula Weight 417.63
Purity >99%
Molecular Weight (g/mol) 417.634
Chemical Name or Material OLDA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.