missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Olaratumab Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558745
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Olaratumab (IMC-3G3) is a fully human IgG1 monoclonal antibody with antitumor activity that selectively binds the external domain of human platelet-derived growth factor receptor (PDGFR)-alpha with high affinity and blocks ligand binding. It is composed of two heavy chain molecule fragments and 2 light chain fragments. Studies show that the treatment of olaratumab in combination with doxorubicin resulted in significant reduction of cancer cell proliferation and tumor growth. Olaratumab was granted accelerated approval (as Lartruvo) as initial therapy to treat adults with certain types of soft tissue sarcoma (STS) in October, 2016.
Specifications
| Olaratumab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| 3G3; IMC-3G3; LY3012207 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25 mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction