missing translation for 'onlineSavingsMsg'
Learn More

Olaparib, MedChemExpress

Product Code. 16474656
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
2 g
Unit Size:
100mg
10mg
1g
200mg
2g
500mg
50mg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16474656 100 mg 100mg
16410819 1 g 1g
16450936 200 mg 200mg
16431536 500 mg 500mg
16449956 50 mg 50mg
16423468 10 mg 10mg
16450056 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 16474656 Supplier Medchem Express Supplier No. HY10162
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Olaparib (AZD2281; KU0059436) is a potent and orally active PARP inhibitor with IC50s of 5 and 1 nM for PARP1 and PARP2, respectively. Olaparib is an autophagy and mitophagy activator.

Olaparib (AZD2281

Specifications

Chemical Name or Material Olaparib
CAS 763113-22-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H23FN4O3
Quantity 100 mg
Synonym AZD2281 KU0059436
Solubility Information DMSO : 100 mg/mL (230.17 mM; Need ultrasonic) ∣DMF : 50 mg/mL (115.09 mM; Need ultrasonic) ∣Ethanol : 3.12 mg/mL (7.18 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(C1=CC(CC(C2=C3C=CC=C2)=NNC3=O)=CC=C1F)N4CCN(C(C5CC5)=O)CC4
Molecular Weight (g/mol) 434.46
Formula Weight 434.46
Percent Purity 96.56%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.