missing translation for 'onlineSavingsMsg'
Learn More

Olaparib, MedChemExpress

Product Code. 16410819
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
2 g
Unit Size:
100mg
10mg
1g
200mg
2g
500mg
50mg
This item is not returnable. View return policy

Product Code. 16410819

Brand: Medchem Express HY10162

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Olaparib (AZD2281; KU0059436) is a potent and orally active PARP inhibitor with IC50s of 5 and 1 nM for PARP1 and PARP2, respectively. Olaparib is an autophagy and mitophagy activator.

Olaparib (AZD2281

Specifications

Chemical Name or Material Olaparib
CAS 763113-22-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H23FN4O3
Quantity 1 g
Synonym AZD2281 KU0059436
Solubility Information DMSO : 100 mg/mL (230.17 mM; Need ultrasonic) ∣DMF : 50 mg/mL (115.09 mM; Need ultrasonic) ∣Ethanol : 3.12 mg/mL (7.18 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(C1=CC(CC(C2=C3C=CC=C2)=NNC3=O)=CC=C1F)N4CCN(C(C5CC5)=O)CC4
Molecular Weight (g/mol) 434.46
Formula Weight 434.46
Percent Purity 96.56%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.