missing translation for 'onlineSavingsMsg'
Learn More

Olanzapine, TRC

Product Code. 18030050
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
Unit Size:
100mg
25mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18030050 500 mg 500mg
18033030 100 mg 100mg
18023130 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18030050 Supplier Toronto Research Chemicals Supplier No. TRCO253750500MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132539-06-1
Molecular Formula C17 H20 N4 S
Molecular Weight (g/mol) 312.43
Synonym 10H-Thieno[2,3-b][1,5]benzodiazepine, 2-methyl-4-(4-methyl-1-piperazinyl)-, 2-Methyl-4-(4-methyl-1-piperazinyl)-10H-thieno[2,3-b][1,5]benzodiazepine, ADAMED, Arenbil, Integrol, LY 170053, Lanzac, Oferta, Olanz, Olanzapine, Oleanz, Olenz, Olimelt, Oliza, Oltal, Prexal, Rexapin, Zolafren, Zolafren Flas, ZypAdhera, Zyprexa, Zyprexa-Velotab, Zyprobiox
IUPAC Name 2-methyl-4-(4-methylpiperazin-1-yl)-10H-thieno[2,3-b][1,5]benzodiazepine
SMILES CN1CCN(CC1)C2=Nc3ccccc3Nc4sc(C)cc24

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C17H20N4S/c1-12-11-13-16(21-9-7-20(2)8-10-21)18-14-5-3-4-6-15(14)19-17(13)22-12/h3-6,11,19H,7-10H2,1-2H3
Formula Weight 312.1409
Percent Purity >95%
Chemical Name or Material Olanzapine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.