missing translation for 'onlineSavingsMsg'
Learn More

Olanzapine Thiolactam Impurity, TRC

Product Code. 18040920
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18040920 5 mg 5mg
18023930 10 mg 10mg
18030530 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18040920 Supplier Toronto Research Chemicals Supplier No. TRCO2537805MG
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1017241-36-9
Molecular Formula C17 H20 N4 O S
Molecular Weight (g/mol) 328.43
Synonym (1Z)-1-[4,5-Dihydro-2-(4-methyl-1-piperazinyl)-4-thioxo-3H-1,5-benzodiazepin-3-ylidene]-2-propanone, (Z)-Olanzapine Ketothiolactam
IUPAC Name (1Z)-1-[4-(4-methylpiperazin-1-yl)-2-sulfanylidene-1H-1,5-benzodiazepin-3-ylidene]propan-2-one
SMILES CN1CCN(CC1)C2=Nc3ccccc3NC(=S)/C/2=C\C(=O)C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C17H20N4OS/c1-12(22)11-13-16(21-9-7-20(2)8-10-21)18-14-5-3-4-6-15(14)19-17(13)23/h3-6,11H,7-10H2,1-2H3,(H,19,23)/b13-11-
Formula Weight 328.1358
Percent Purity >95%
Chemical Name or Material (Z)-Olanzapine Ketothiolactam
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.