missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olanzapine , MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
Description
Olanzapine (LY170053) is a selective, orally active monoaminergic antagonist with high affinity binding to serotonin H1, 5HT2A/2C, 5HT3, 5HT6 (Ki=7, 4, 11, 57, and 5 nM, respectively), dopamine D1-4 (Ki=11 to 31 nM), muscarinic M1-5 (Ki=1.9-25 nM), and adrenergic α1 receptor (Ki=19 nM). Olanzapine is an atypical antipsychotic.
Specifications
Specifications
| Chemical Name or Material | Olanzapine |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C17H20N4S |
| Quantity | 100 mg |
| Formula Weight | 312.43 |
| Grade | Research |
| Physical Form | Solid |
| For Use With (Application) | Cancer-programmed cell death |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction