missing translation for 'onlineSavingsMsg'
Learn More

Olanzapine, 1mg/ml in Acetonitrile, Lipomed

Product Code. 30460767
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460767 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30460767 Supplier Lipomed Supplier No. LPMOLZ1064FB1LA
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Olanzapine, 1mg/ml in Acetonitrile
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Olanzapine
CAS 132539-06-1
Analyte or Component Names Olanzapine
Shipping Condition Room Temperature
Molecular Formula C17 H20 N4 S
Concentration 1 mg/mL
Synonym Olanzapine, 2-Methyl-4-(4-methyl-1-piperazinyl)-10 H-thieno[2,3-b][1,5 ]benzodiazepine, LY 170053, Lanzac, Zyprexa
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES CN1CCN(CC1)C2=Nc3ccccc3Nc4sc(C)cc24
IUPAC Name 2-methyl-4-(4-methylpiperazin-1-yl)-10 H-thieno[2,3-b][1,5 ]benzodiazepine
Solvent or Matrix Acetonitrile
Industry Type Pharmaceutical
Molecular Weight (g/mol) 312.43
Quantity 1 mL
Formula Weight 312.1409 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.