missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OLAH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558179
This item is not returnable.
View return policy
Description
Immunogen sequence: AEAKEFVKQC SPIIRADLNI VRSCTSNVPS KAVLSCDLTC FVGSEDIAKD MEAWKDVTSG NAKIYQLPGG HFYLLD Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 43%.
Specifications
| OLAH | |
| Polyclonal | |
| Unconjugated | |
| OLAH | |
| Augmented in rheumatoid arthritis 1; AURA1; OLAH; Oleoyl-ACP hydrolase; oleoyl-acyl-carrier-protein hydrolase; S-acyl fatty acid synthase thioesterase, medium chain; SAST; TE2; THEDC1; Thioesterase 2; thioesterase domain containing 1; thioesterase domain-containing protein 1; Thioesterase II | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55301 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NV23 | |
| OLAH | |
| Recombinant protein corresponding to Human OLAH. Recombinant protein control fragment (Product #RP-97121). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction