missing translation for 'onlineSavingsMsg'
Learn More

Euro Protection™ Okenite Safety Shoes

Product Code. 12064295
Change view
Click to view available options
Size:
37
38
39
41
42
44
45
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12064295 42 Each
12014295 37 Each
12024295 38 Each
12034295 39 Each
12054295 41 Each
12084295 44 Each
12094295 45 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 12064295 Supplier Euro Protection™ Supplier No. 9OKEL42
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Euro Protection™ Okenite Safety Shoes are breathable, easy to put on/take off, suitable for food industry use, and feature a composite toe cap for protection.

  • Suitable for food industry use
  • Easy-on and easy-off
  • Protective cover
  • Rounded heel for more comfortable driving
  • Microfiber upper
  • Breathable 3D mesh lining
  • Removable, textile and foam insole
  • Composite toe cap
  • Injected polyurethane midsole
  • Polyurethane outsole

Specifications

Color White, Gray
For Use With (Application) Suitable for food industry
Heel Style Rounded
Liner Material Breathable 3D mesh lining
Material Microfiber
Size 42
Sole Features Removable, textile and foam insole, injected polyurethane midsole, polyurethane outsole.
Sole Material Polyurethane
Style Low-cut
Toe Material Composite
Weight (Metric) 427 g per shoe in size 8
Certifications/Compliance (Europe) EN ISO 20345 : 2011
Product Type Safety Shoe
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.