missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Okadaic acid

Product Code. 18763474
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18763474 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18763474 Supplier Tocris Bioscience™ Supplier No. 1136/100U

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Protein phosphatase 1 and 2A inhibitor

Okadaic acid is a potent inhibitor of protein phosphatase 1 (IC50 = 3 nM) and protein phosphatase 2A (IC50 = 0.2-1 nM). Displays > 100,000,000-fold selectivity over PP2B and PP2C. Tumor promoter. Shown to activate atypical protein kinase C in adipocytes.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 9,10-Deepithio-9,10-didehydroacanthifolicin
CAS 78111-17-8
Quantity 100 μg
Target Protein Ser/Thr Phosphatase Inhibitors
Molecular Formula C44H68O13
Purity 0.9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.