Learn More
Kern Sohn OIML Class M1 Flat Sheet Test Weight
Click to view available options
Weight (Metric):
1 mg
2 mg
5 mg
10 mg
20 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
Each
Description
- Flat polygonal sheet
- Polished stainless steel
Specifications
Specifications
| Classification | M1 |
| Material | Stainless Steel |
| Weight (Metric) | 5 mg |
| Tolerance | ± 0.2 mg |