Learn More
Kern Sohn OIML Class E1 Knob Test Weight
Product Code. 17311853
Encompass_Preferred
Click to view available options
Weight (Metric):
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
20 kg
50 kg
Unit Size:
Each
Product Code. 17311853
Brand: Kern Sohn 30701
Only null left
Add to basket
Add to basket
Single test weight constructed of stainless steel.
  • Knob shape
  • Polished stainless steel
Specifications
Classification E1
Material Stainless Steel
Weight (Metric) 1 g
Tolerance ± 0.01 mg