Learn More
OHAUS™ Sample Pan for MB Series Moisture Analyzers
Product Code. p-3207089 Shop All Ohaus Products
Click to view available options
Type:
Disposable Sample Pan
Reusable Sample Pan
Quantity:
3/Pk.
80/Pk.
Unit Size:
Pack of 3
Pack of 80
Product Code. 11804242
Brand: OHAUS™ 80850086
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Sample Pans for OHAUS™ MB series moisture analyzers.
  • Shape: round
  • Choose disposable or reusable
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) US: MB35,MB45,MB27,MB90,MB120 Moisture Analyzers
EU: MB27, MB25, MB90-GB, MB120-GB, MB23, MB120-EU
Size 101.6 mm diameter
Quantity 80/Pk.
Type Disposable Sample Pan