missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OGT Recombinant Rabbit Monoclonal Antibody (JB44-39)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA534661

Product Code. 17135333

  • £398.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive Control: Hela, SiHa, A431, PC-3M, rat epididymis tissue, human liver tissue, human kidney tissue, mouse testis tissue.

This gene encodes a glycosyltransferase that catalyzes the addition of a single N-acetylglucosamine in O-glycosidic linkage to serine or threonine residues. Since both phosphorylation and glycosylation compete for similar serine or threonine residues, the two processes may compete for sites, or they may alter the substrate specificity of nearby sites by steric or electrostatic effects. The protein contains multiple tetratricopeptide repeats that are required for optimal recognition of substrates. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

OGT
Recombinant Monoclonal
1 mg/mL
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
O15294
OGT
Synthetic peptide within Human OGT aa 997-1,046.
100 μL
Primary
Human
Antibody
IgG
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
JB44-39
Unconjugated
OGT
1110038P24Rik; 4831420N21Rik; AI115525; FLJ23071; HINCUT-1; HRNT1; MGC22921; O linked N-acetylglucosamine transferase; O-GLCNAC; o-glcnac transferase; O-GlcNAc transferase p110 subunit; O-GlcNAc transferase subunit p110; OGT; Ogtl; O-linked N-acetylglucosamine (GlcNAc) transferase; O-linked N-acetylglucosamine (GlcNAc) transferase (UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase); O-linked N-acetylglucosamine transferase; O-linked N-acetylglucosamine transferase 110 kDa subunit; UDP-N-acetylglucosamine:polypeptide-N- acetylglucosaminyl transferase; UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase; UDP-N-acetylglucosamine--peptide N-acetylglucosaminyltransferase 110 kDa subunit; uridinediphospho-N-acetylglucosamine:polypeptide beta-N-acetylglucosaminyl transferase
Rabbit
Protein A
RUO
8473
-20°C, Avoid Freeze/Thaw Cycles, store in dark
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.