missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ OGR1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£179.00 - £651.00

Specifications

Antigen OGR1
Dilution Western Blot : 5-10 μg/mL, Immunohistochemistry, Functional
Applications Immunohistochemistry, Western Blot, Functional Assay
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30066946
View Documents
Novus Biologicals™
NBP3-30855-25ug
25 μg
£179.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30067206
View Documents
Novus Biologicals™
NBP3-30855-100ug
100 μg
£651.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

OGR1 Polyclonal antibody specifically detects OGR1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Functional Assay
Specifications

Specifications

OGR1
Immunohistochemistry, Western Blot, Functional Assay
Unconjugated
Rabbit
GPCR
Phosphate buffered saline
8111
IgG
Protein A or G purified
Western Blot : 5-10 μg/mL, Immunohistochemistry, Functional
Polyclonal
Purified
RUO
Human
G protein-coupled receptor 68, GPR12A, G-protein coupled receptor 68, OGR-1, ovarian cancer G protein-coupled receptor, 1, ovarian cancer G-protein coupled receptor 1, Sphingosylphosphorylcholine receptor
Rabbits were immunized with recombinant protein derived from human OGR1 (GPR68).
Primary
Store at -20°C. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.