missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Oglufanide

Product Code. 16465818
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16465818 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16465818 Supplier MedChemExpress Supplier No. HY13718
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Oglufanide (H-Glu-Trp-OH) is a dipeptide immunomodulator isolated from calf thymus. Oglufanide inhibits vascular endothelial growth factor (VEGF). Oglufanide can stimulate the immune response to hepatitic C virus (HCV) and intracellular bacterial infections.

Specifications

Purity 99.49%
CAS 38101-59-6
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
For Use With (Application) COVID-19-anti-virus
Molecular Weight (g/mol) 333.34 g/mol
Product Type Natural Products
Quantity 10 mg
Shipping Condition Room temperature
Solubility DMSO : 15.5 mg/mL (46.50 mM∣Need ultrasonic and warming)
Synonym H-Glu-Trp-OH∣ L-Glutamyl-L-tryptophan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.